Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platform for Eppendorf
®
New Brunswick
™
Innova
®
2100/40/40R/42/42R & Excella
®
E24/E24R Shakers
Products
Manufacturer
Length (cm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EPM1194-9908
Platform for Eppendorf
®
New Brunswick
™
Innova
®
2100/40/40R/42/42R & Excella
®
E24/E24R Shakers,
for 2 L Erlenmeyer flasks, dedicated, phenolic resin
Expand
View Pricing
EPM1194-9904
Platform for Eppendorf
®
New Brunswick
™
Innova
®
2100/40/40R/42/42R & Excella
®
E24/E24R Shakers,
for 125 mL Erlenmeyer flasks, dedicated, phenolic resin
Expand
View Pricing
EPM1194-9905
Platform for Eppendorf
®
New Brunswick
™
Innova
®
2100/40/40R/42/42R & Excella
®
E24/E24R Shakers,
for 250 mL Erlenmeyer flasks, dedicated, phenolic resin
Expand
View Pricing
EPM1194-9903
Platform for Eppendorf
®
New Brunswick
™
Innova
®
2100/40/40R/42/42R & Excella
®
E24/E24R Shakers,
for 50 mL Erlenmeyer flasks, non-Interchangeable, phenolic resin
Expand
View Pricing
EPM1194-9906
Platform for Eppendorf
®
New Brunswick
™
Innova
®
2100/40/40R/42/42R & Excella
®
E24/E24R Shakers,
for 500 mL Erlenmeyer flasks, dedicated, phenolic resin
Expand
View Pricing
EPM1194-9907
Platform for Eppendorf
®
New Brunswick
™
Innova
®
2100/40/40R/42/42R & Excella
®
E24/E24R Shakers,
for 1 L Erlenmeyer flasks, dedicated, phenolic resin
Expand
View Pricing