Skip to Content
Merck
Piroxicam European Pharmacopoeia (EP) Reference Standard

Piroxicam

Piroxicam European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-Hydroxy-2-methyl-3-(pyrid-2-yl-carbamoyl)-2H-1,2-benzothiazine 1,1-dioxide, Piroxicam
Empirical Formula (Hill Notation):
C15H13N3O4S
CAS Number:
Molecular Weight:
331.35

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique