Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette Stand
Products
Manufacturer
Height (mm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
HS206204
Pipette Stand,
acrylic, to hold, 4 pipette(s), clear
Expand
View Pricing
HS20613C
Pipette Stand,
acrylic, to hold, 6 pipette(s), blue
Expand
View Pricing
HS206203
Pipette Stand,
acrylic, to hold, 3 pipette(s), clear
Expand
View Pricing
HS20613E
Pipette Stand,
acrylonitrile-butadiene-styrene (ABS), to hold, 6 pipette(s), blue
Expand
View Pricing
HS20620
Pipette Stand,
acrylic, to hold, 6 pipette(s), clear
Expand
View Pricing