Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette bulb
Products
Brand
Sigma-Aldrich (3)
Capacity (mL)
Volume (ml)
Color
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z136042
Pipette bulb,
capacity 2 mL
Expand
View Pricing
Z509035
Pipette bulb,
volume 1 mL
Expand
View Pricing
Z186678
Pipette bulb,
capacity 5 mL
Expand
View Pricing
Z509043
Pipette bulb,
volume 3 mL
Expand
View Pricing
Z136069
Pipette bulb,
capacity 60 mL
Expand
View Pricing
Z136050
Pipette bulb,
capacity 8 mL
Expand
View Pricing
Z186708
Pipette bulb,
capacity 150 mL
Expand
View Pricing
Z186694
Pipette bulb,
capacity 35 mL
Expand
View Pricing
Z186686
Pipette bulb,
capacity 15 mL
Expand
View Pricing