
Phenyltoloxamine citrate

Synonym(s):
Phenyltoloxamine citrate salt, N,N-Dimethyl-2([α-phenyl-o-tolyl]oxy)ethylamine
Empirical Formula (Hill Notation):
C17H21NO · C6H8O7
CAS Number:
Molecular Weight:
447.48
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2