Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phenolphthalein solution
Empirical Formula (Hill Notation):
C
20
H
14
O
4
CAS Number:
77-09-8
Molecular Weight:
318.32
Products
Product Category
dyes (1)
Brand
Sigma-Aldrich (4)
Color
Formula Weight
pH Value
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
319236
Phenolphthalein solution,
0.5 wt. % in ethanol: water (1:1)
Expand
View Pricing
40-1022
Phenolphthalein solution,
1 % (w/v), pH 7.8-10.0
Expand
View Pricing
40-1023
Phenolphthalein solution,
0.1 % (w/v), pH 7.8-10.0
Expand
View Pricing
40-2031
Phenolphthalein solution,
0.04 % (w/v), pH 7.8-10.0
Expand
View Pricing