Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Penicillin G sodium salt
Synonym(s):
Benzylpenicillin sodium salt
Empirical Formula (Hill Notation):
C
16
H
17
N
2
NaO
4
S
CAS Number:
69-57-8
Molecular Weight:
356.37
EC Number:
200-710-2
Products
Product Category
antibiotics (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Quality Segment
Mode of Action
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
P3032
Penicillin G sodium salt,
powder, suitable for cell culture, BioReagent
Expand
View Pricing
13752
Penicillin G sodium salt,
96.0-102.0%
Expand
View Pricing
PENNA
Penicillin G sodium salt,
~1650 U/mg
Expand
View Pricing