Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pectinase from
Aspergillus niger
Synonym(s):
Poly-(1,4-α-D-galacturonide) glycanohydrolase, Polygalacturonase solution from
Aspergillus niger
, Polygalacturonase
CAS Number:
9032-75-1
EC Number:
232-885-6
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (2)
Sustainability
Biological Source
Color
Feature
Physical Form
Product Line
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P4716
Pectinase from
Aspergillus niger
,
BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Expand
View Pricing
17389
Pectinase from
Aspergillus niger
,
powder, slightly beige, >1 U/mg
Expand
View Pricing