
Panthenol, racemic

Synonym(s):
DL-Panthenol, (±)-α,γ-Dihydroxy-N-(3-hydroxypropyl)-β,β-dimethylbutyramide, (±)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, DL-Pantothenyl alcohol
Linear Formula:
HOCH2C(CH3)2CH(OH)CONH(CH2)3OH
CAS Number:
Molecular Weight:
205.25
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing