Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium/Silver
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF02839822
Palladium/Silver,
foil, Pd 75%/Ag 25%, thickness 0.01 mm, size 50 × 50 mm
Expand
View Pricing
GF45621671
Palladium/Silver,
wire, Pd 75%/Ag 25%, 1.0 mm diameter, length 0.2 m, temper as drawn
Expand
View Pricing
GF32069561
Palladium/Silver,
foil, Pd 75%/Ag 25%, thickness 0.01 mm, size 25 × 25 mm
Expand
View Pricing
GF50504554
Palladium/Silver,
wire, Pd 75%/Ag 25%, 1.0 mm diameter, length 0.1 m, temper as drawn
Expand
View Pricing
GF61228290
Palladium/Silver,
wire, 1.0 mm diameter, Pd75%/Ag25%, length 0.05 m, temper as drawn
Expand
View Pricing
GF85001150
Palladium/Silver,
wire, 1.0 mm diameter, Pd75%/Ag25%, length 0.5 m, temper as drawn
Expand
View Pricing