
Ouabain

Synonym(s):
Ouabain octahydrate, 1β,3β,5β,11α,14,19-Hexahydroxycard-20(22)-enolide 3-(6-deoxy-α-L-mannopyranoside), Acocantherine, G-Strophanthin
Empirical Formula (Hill Notation):
C29H44O12 · 8H2O
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing