
Orphenadrine Citrate

Synonym(s):
Orphenadrine citrate salt, β-Dimethylaminoethyl 2-methylbenzhydryl ether citrate salt, o-Methyldiphenhydramine citrate salt
Empirical Formula (Hill Notation):
C18H23NO · C6H8O7
CAS Number:
Molecular Weight:
461.50
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2