Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Orange II sodium salt
Synonym(s):
4-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid sodium salt, Acid Orange 7, Acid Orange A, Orange II, Tropaeolin 000 No. 2
Empirical Formula (Hill Notation):
C
16
H
11
N
2
NaO
4
S
CAS Number:
633-96-5
Molecular Weight:
350.32
EC Number:
211-199-0
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Physical Form
Product Line
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
195235
Orange II sodium salt,
(Certified by the Biological Stain Commission), Dye content ≥85 %
Expand
View Pricing
69143
Orange II sodium salt,
analytical standard
Expand
View Pricing