Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Scout
®
SPX Balance
Products
Product Category
balances (12)
Manufacturer
Diameter (mm)
Weighing Capacity (g)
Resolution (g)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z742224
Ohaus
®
Scout
®
SPX Balance,
Model SPX2202, weighing capacity 2200 g, resolution: 0.01 g
Expand
View Pricing
Z742229
Ohaus
®
Scout
®
SPX Balance,
Model SPX6201, weighing capacity 6200 g, resolution: 0.1 g
Expand
View Pricing
Z742220
Ohaus
®
Scout
®
SPX Balance,
Model SPX222, weighing capacity 220 g, resolution: 0.01 g
Expand
View Pricing
Z742227
Ohaus
®
Scout
®
SPX Balance,
Model SPX2201, weighing capacity 2200 g, resolution: 0.1 g
Expand
View Pricing
Z742222
Ohaus
®
Scout
®
SPX Balance,
Model SPX622, weighing capacity 620 g, resolution: 0.01 g
Expand
View Pricing
Z742219
Ohaus
®
Scout
®
SPX Balance,
Model SPX223, weighing capacity 220 g, resolution: 0.001 g
Expand
View Pricing
Z742218
Ohaus
®
Scout
®
SPX Balance,
Model SPX123, weighing capacity 120 g, resolution: 0.001 g
Expand
View Pricing
Z742225
Ohaus
®
Scout
®
SPX Balance,
Model SPX421, weighing capacity 420 g, resolution: 0.1 g
Expand
View Pricing
Z742226
Ohaus
®
Scout
®
SPX Balance,
Model SPX621, weighing capacity 620 g, resolution: 0.1 g
Expand
View Pricing
Z742221
Ohaus
®
Scout
®
SPX Balance,
Model SPX422, weighing capacity 420 g, resolution: 0.01 g
Expand
View Pricing
Z742223
Ohaus
®
Scout
®
SPX Balance,
Model SPX1202, weighing capacity 1200 g, resolution: 0.01 g
Expand
View Pricing
Z742230
Ohaus
®
Scout
®
SPX Balance,
Model SPX8200, weighing capacity 8200 g, resolution: 1 g
Expand
View Pricing