
Octane-1-sulfonic acid sodium salt

Synonym(s):
Octane-1-sulfonic acid sodium salt
Empirical Formula (Hill Notation):
C8H17NaO3S
CAS Number:
Molecular Weight:
216.27
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing