Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Octane
Synonym(s):
n
-Octane
Linear Formula:
CH
3
(CH
2
)
6
CH
3
CAS Number:
111-65-9
Molecular Weight:
114.23
EC Number:
203-892-1
Products
Product Category
solvents (2)
building blocks (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Anhydrous (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
412236
Octane,
reagent grade, 98%
Expand
View Pricing
296988
Octane,
anhydrous, ≥99%
Expand
View Pricing
657042
Octane,
electronic grade, ≥99.999% metals basis, ≥99% (CP)
Expand
View Pricing
74820
Octane,
analytical standard
Expand
View Pricing