Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
UpCell
™
Surface cell culture multidish
Products
Product Category
plates (4)
dishes (1)
Manufacturer
Length (mm)
Surface Area cm²
Wells
Working Volume (ml)
Color
Plate Material
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z694347
Nunc
®
UpCell
™
Surface cell culture multidish,
size 96 wells, sterile, surface area 0.33 cm
2
Expand
View Pricing
Z688789
Nunc
®
UpCell
™
Surface cell culture multidish,
Expand
View Pricing
Z688800
Nunc
®
UpCell
™
Surface cell culture multidish,
Expand
View Pricing
Z688797
Nunc
®
UpCell
™
Surface cell culture multidish,
Expand
View Pricing
Z688770
Nunc
®
UpCell
™
Surface cell culture multidish,
Expand
View Pricing