Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
Lab-Tek
®
II - CC
2
™
Chamber Slide
™
system
Products
Product Category
slides (3)
Brand
Sigma-Aldrich (2)
Manufacturer
Surface Area cm²
Working Volume (ml)
Color
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S6815
Nunc
®
Lab-Tek
®
II - CC
2
™
Chamber Slide
™
system,
8 wells, glass slide, 0.7 cm
2
/well, sterile
Expand
View Pricing
S6690
Nunc
®
Lab-Tek
®
II - CC
2
™
Chamber Slide
™
system,
4 wells, glass slide, 1.7 cm
2
/well, sterile
Expand
View Pricing
S6565
Nunc
®
Lab-Tek
®
II - CC
2
™
Chamber Slide
™
system,
2 wells, glass slide, 4.0 cm
2
/well, sterile
Expand
View Pricing