Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
inoculating loops
Products
Product Category
loops (4)
Brand
Sigma-Aldrich (3)
Color
Manufacturer
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
I7523
Nunc
®
inoculating loops,
blue, size 10 μL, sleeve of 50
Expand
View Pricing
I7898
Nunc
®
inoculating loops,
blue, pkg of 600 (50 sleeves of 12 each), size 10 μL
Expand
View Pricing
I7773
Nunc
®
inoculating loops,
colorless, size 1 μL, sleeve of 50
Expand
View Pricing
I7648
Nunc
®
inoculating loops,
size 1 μL, pkg of 600 (50 sleeves of 12 each)
Expand
View Pricing