Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nitinol
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GF15396419
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 1 m, condition shape-memory alloy
Expand
View Pricing
GF84827537
Nitinol,
foil, thickness 0.5 mm, Ni55%/Ti45%, size 90 × 90 mm, temper annealed
Expand
View Pricing
GF24053677
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 10 m, condition shape-memory alloy
Expand
View Pricing
GF65520727
Nitinol,
rod, Ni55%/Ti45%, 2.5 mm diameter, length 100 mm, condition superelastic
Expand
View Pricing
GF72230554
Nitinol,
rod, 2.5 mm diameter, Ni55%/Ti45%, length 500 mm, condition superelastic
Expand
View Pricing
GF55626778
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 0.5 m, condition shape-memory alloy
Expand
View Pricing
GF25681035
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 0.5 m, condition shape-memory alloy
Expand
View Pricing
GF50859524
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 0.5 m, condition shape-memory alloy
Expand
View Pricing
GF32583179
Nitinol,
foil, Ni55%/Ti 45%, thickness 0.5 mm, size 45 × 45 mm, temper annealed
Expand
View Pricing
GF93676168
Nitinol,
wire, 0.125 mm diameter, Ni55%/Ti45%, length 2 m, condition shape-memory alloy
Expand
View Pricing
GF62412319
Nitinol,
rod, 2.5 mm diameter, Ni55%/Ti45%, length 1000 mm, condition superelastic
Expand
View Pricing
GF82647513
Nitinol,
wire, Ni55%/Ti45%, 0.125 mm diameter, length 2 m, condition shape-memory alloy
Expand
View Pricing
GF31623005
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 5 m, condition shape-memory alloy
Expand
View Pricing
GF49974643
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 1 m, condition shape-memory alloy
Expand
View Pricing
GF53826475
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 1 m, condition shape-memory alloy
Expand
View Pricing
GF18215209
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 2 m, condition shape-memory alloy
Expand
View Pricing
GF18353606
Nitinol,
wire, Ni55%/Ti 45%, 0.125 mm diameter, length 5 m, condition shape-memory alloy
Expand
View Pricing