Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ninhydrin
Synonym(s):
Ninhydrin, 1,2,3-Indantrione monohydrate, 2,2-Dihydroxy-1,3-indanedione, Trioxohydrindene monohydrate
Empirical Formula (Hill Notation):
C
9
H
6
O
4
CAS Number:
485-47-2
Molecular Weight:
178.14
EC Number:
207-618-1
Products
Product Category
reagents (2)
building blocks (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.06762
Ninhydrin,
GR for analysis ACS,Reag. Ph Eur
Expand
View Pricing
151173
Ninhydrin,
ACS reagent
Expand
View Pricing
NX0403
Ninhydrin,
Expand
View Pricing
N4876
Ninhydrin,
suitable for amino acid detection
Expand
View Pricing
21-1510
Ninhydrin,
SAJ special grade, ≥98.0%
Expand
View Pricing