
Nimbolide

Synonym(s):
(Methyl (2aR,5aR,6S,6aR,8R,9aR,10aS,10bR,10cR)-8-(furan-3-yl)-(2a,5a,6,6a,8,9,9a,10a,10b,10c)- decahydro-2a,5a,6a,7-tetramethyl-2,5-dioxo-2H,5H-cyclopenta[d′]naphtho[1,8-bc:2,3-b′]difuran-6- acetate)
Empirical Formula (Hill Notation):
C27H30O7
CAS Number:
Molecular Weight:
466.52
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Special Grade
Showing 1-3 of 3