Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nile Red
Synonym(s):
Nile Blue A Oxazone
Empirical Formula (Hill Notation):
C
20
H
18
N
2
O
2
CAS Number:
7385-67-3
Molecular Weight:
318.37
EC Number:
230-966-0
Products
Product Category
detergents (1)
dyes (1)
stains (1)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
72485
Nile Red,
for microscopy
Expand
View Pricing
19123
Nile Red,
BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Expand
View Pricing
N3013
Nile Red,
Powder
Expand
View Pricing