
Nifedipine impurity A

Synonym(s):
Oxidized Nifedipine, 2,6-Dimethyl-4-(2´-nitrophenyl)-3,5-pyridinecarboxylic acid dimethyl ester
Empirical Formula (Hill Notation):
C17H16N2O6
CAS Number:
Molecular Weight:
344.32
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing