Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Niclosamide
Synonym(s):
2′,5-Dichloro-4′-nitrosalicylanilide
Empirical Formula (Hill Notation):
C
13
H
8
Cl
2
N
2
O
4
CAS Number:
50-65-7
Molecular Weight:
327.12
EC Number:
200-056-8
Products
Product Category
antibiotics (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
N3510
Niclosamide,
Expand
View Pricing