Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel microhardness test block (Knoop)
Products
Manufacturer
Physical Form
Quality Segment
Reference Material Type
Special Grade
certified reference material (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
NIST1905
Nickel microhardness test block (Knoop),
Expand
View Pricing
NIST1906
Nickel microhardness test block (Knoop),
Expand
View Pricing
NIST1907
Nickel microhardness test block (Knoop),
Expand
View Pricing
NIST1895
Nickel microhardness test block (Knoop),
NIST
®
SRM
®
1895
Expand
View Pricing