Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Neon-
20
Ne
Linear Formula:
20
Ne
Molecular Weight:
19.99
Products
Product Category
gases (4)
stable isotopes (4)
Brand
Sigma-Aldrich (4)
Formula Weight
Physical Form
Purity
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
601667
Neon-
20
Ne,
99.95 atom %, 99.995% (CP)
Expand
View Pricing
601691
Neon-
20
Ne,
99.9 atom %
Expand
View Pricing
601675
Neon-
20
Ne,
70 atom %
Expand
View Pricing
601683
Neon-
20
Ne,
99.95 atom %
Expand
View Pricing