Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Neodymium-iron-boron alloy, Nd2Fe14B
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF29925861
Neodymium-iron-boron alloy, Nd2Fe14B,
sheet, 5 x 5mm, 2mm thickness, rare earth magnet, parylene coating
Expand
View Pricing
GF40733412
Neodymium-iron-boron alloy, Nd2Fe14B,
sheet, 15 x 15mm, 5mm thickness, rare earth magnet, uncoated coating
Expand
View Pricing
GF51579995
Neodymium-iron-boron alloy, Nd2Fe14B,
15mm rod, 6.35mm diameter, rare earth magnet, uncoated coating
Expand
View Pricing
GF53111416
Neodymium-iron-boron alloy, Nd2Fe14B,
3mm rod, 15mm diameter, rare earth magnet, uncoated coating
Expand
View Pricing
GF91676997
Neodymium-iron-boron alloy, Nd2Fe14B,
2mm rod, 6.35mm diameter, rare earth magnet, parylene coating
Expand
View Pricing