Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Near-IR Fluorescent PLGA nanoparticles
Products
Brand
Sigma-Aldrich (11)
Physical Form
Particle Size (µm)
Polymer Composition
Polymer Type
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
LGFN100
Near-IR Fluorescent PLGA nanoparticles,
100 nm average diameter
Expand
View Pricing
LGFN200
Near-IR Fluorescent PLGA nanoparticles,
200 nm average diameter
Expand
View Pricing
LGFN10K
Near-IR Fluorescent PLGA nanoparticles,
10 μm average diameter
Expand
View Pricing
LGFN500
Near-IR Fluorescent PLGA nanoparticles,
500 nm average diameter
Expand
View Pricing
LGFN1000
Near-IR Fluorescent PLGA nanoparticles,
1 μm average diameter
Expand
View Pricing
LGFN50K
Near-IR Fluorescent PLGA nanoparticles,
50 μm average diameter
Expand
View Pricing
LGFN40K
Near-IR Fluorescent PLGA nanoparticles,
40 μm average diameter
Expand
View Pricing
LGFN2000
Near-IR Fluorescent PLGA nanoparticles,
2 μm average diameter
Expand
View Pricing
LGFN20K
Near-IR Fluorescent PLGA nanoparticles,
20 μm average diameter
Expand
View Pricing
LGFN5000
Near-IR Fluorescent PLGA nanoparticles,
5 μm average diameter
Expand
View Pricing
LGFN30K
Near-IR Fluorescent PLGA nanoparticles,
30 μm average diameter
Expand
View Pricing