Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
Labtop cooler
Products
Brand
Sigma-Aldrich (4)
Color
Manufacturer
Height (mm)
Length (mm)
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
C2437
Nalgene
®
Labtop cooler,
−20°C Labtop cooler, Jr., transparent lid
Expand
View Pricing
C9985
Nalgene
®
Labtop cooler,
-20 °C labtop cooler, polycarbonate, blue, pack of 1
Expand
View Pricing
C2312
Nalgene
®
Labtop cooler,
−20°C Labtop cooler, gel-filled lid
Expand
View Pricing
Z359084
Nalgene
®
Labtop cooler,
0°C Labtop cooler, gel-filled lid
Expand
View Pricing
Z370509
Nalgene
®
Labtop cooler,
0°C Labtop cooler, clear lid
Expand
View Pricing
Z359076
Nalgene
®
Labtop cooler,
0°C Labtop cooler, Jr., transparent lid
Expand
View Pricing
Z370517
Nalgene
®
Labtop cooler,
0°C Labtop cooler, clear lid
Expand
View Pricing