Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
laboratory notebooks
Products
Product Category
books (4)
Brand
Sigma-Aldrich (2)
Color
Manufacturer
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z511676
Nalgene
®
laboratory notebooks,
standard paper pages, lined (5 mm spacing)
Expand
View Pricing
Z261009
Nalgene
®
laboratory notebooks,
standard paper pages, lined (3 lines per in.)
Expand
View Pricing
Z511668
Nalgene
®
laboratory notebooks,
standard paper pages, 5 mm grid
Expand
View Pricing
Z135569
Nalgene
®
laboratory notebooks,
PE cover,
1
/
4
in. grid
Expand
View Pricing