Skip to Content
Merck
Myristic-d27 acid 98 atom % D, 99% (CP)

Myristic-d27 acid

Myristic-d27 acid 98 atom % D, 99% (CP)
Synonym(s):
Tetradecanoic-d27 acid
Linear Formula:
CD3(CD2)12CO2H
CAS Number:
Molecular Weight:
255.54

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing