Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mutanolysin from
Streptomyces globisporus
ATCC 21553
CAS Number:
55466-22-3
Products
Product Category
enzymes (4)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Quality Segment
Application
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
M9901
Mutanolysin from
Streptomyces globisporus
ATCC 21553,
lyophilized powder, ≥4000 units/mg protein (biuret), Chromatographically purified
Expand
View Pricing
SAE0092
Mutanolysin from
Streptomyces globisporus
ATCC 21553,
free of DNA contaminants, suitable for Microbiome research, lyophilized powder, ≥4000 units/mg protein (biuret)
Expand
View Pricing
SRE0007
Mutanolysin from
Streptomyces globisporus
ATCC 21553,
Suitable for manufacturing of diagnostic kits and reagents, ≥4000 units/mg protein (biuret)
Expand
View Pricing
M4782
Mutanolysin from
Streptomyces globisporus
ATCC 21553,
0.2 μm filtered, lyophilized powder, ≥4000 units/mg protein (biuret)
Expand
View Pricing