Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Murexide
Synonym(s):
Murexide, 5,5′-Nitrilodibarbituric acid monoammonium salt, Ammonium purpurate
Empirical Formula (Hill Notation):
C
8
H
8
N
6
O
6
CAS Number:
3051-09-0
Molecular Weight:
284.19
EC Number:
221-266-6
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.06161
Murexide,
metal indicator, ACS
Expand
View Pricing
222461
Murexide,
ACS reagent
Expand
View Pricing