Skip to Content
Merck
Multicomponent Alcohol Mix 1000 1000 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®

Multicomponent Alcohol Mix 1000

Multicomponent Alcohol Mix 1000 1000 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®
Synonym(s):
Multi-component Alcohol Mix

Products

Product Category
Brand
Manufacturer
Physical Form
Format
Chemical Composition
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique