Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Culture Tubes
Products
Product Category
tubes (9)
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
MTCT8225
MTC
™
Bio Culture Tubes,
capacity 14 mL, polystyrene, sterile, pack of 1000 ea (40 x bag 25EA)
Expand
View Pricing
MTCT8235
MTC
™
Bio Culture Tubes,
capacity 14 mL, polypropylene, sterile, pack of 1000 ea (40 x bag 25EA)
Expand
View Pricing
MTCT8305
MTC
™
Bio Culture Tubes,
capacity 5 mL, polystyrene, sterile, pack of 1000 ea (40 x bag 25EA)
Expand
View Pricing
MTCT8250
MTC
™
Bio Culture Tubes,
capacity 14 mL, polystyrene, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCT8260
MTC
™
Bio Culture Tubes,
capacity 14 mL, polypropylene, sterile, pack of 500 ea (individually wrapped)
Expand
View Pricing
MTCT8301
MTC
™
Bio Culture Tubes,
capacity 5 mL, polypropylene, sterile, pack of 1000 ea (40 x bag 25EA)
Expand
View Pricing
MTCT8320
MTC
™
Bio Culture Tubes,
capacity 5 mL, polypropylene, sterile, pack of 1000 ea (8 x bag 125EA)
Expand
View Pricing
MTCT8300
MTC
™
Bio Culture Tubes,
capacity 5 mL, polystyrene, sterile, pack of 1000 ea (8 x bag 125EA)
Expand
View Pricing
MTCC25957
MTC
™
Bio Culture Tubes,
capacity 7 mL, conical bottom, non-sterile, pack of 200 ea
Expand
View Pricing