
Monoclonal Anti-PRAME antibody produced in mouse

Synonym(s):
CT130, MAPE
Products
Product Category
Brand
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing