Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molecular Sieve, 5Å
Products
Product Category
sieves (3)
columns (1)
Brand
Supelco (4)
Physical Form
Agency
Quality Segment
Package Size
Pore Size (Å)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
20302
Molecular Sieve, 5Å,
60-80 mesh, bottle of 50 g
Expand
View Pricing
20301
Molecular Sieve, 5Å,
45-60 mesh, bottle of 50 g
Expand
View Pricing
20300
Molecular Sieve, 5Å,
30-40 mesh, bottle of 50 g
Expand
View Pricing
20298
Molecular Sieve, 5Å,
water vapor trap refill, volume ~460 cc
Expand
View Pricing