Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mini-Centrifuge with 2 rotors
Products
Product Category
centrifuges (6)
Color
Manufacturer
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z742582
Mini-Centrifuge with 2 rotors,
blue, AC/DC input 115 V AC, US 3-pin plug
Expand
View Pricing
Z742583
Mini-Centrifuge with 2 rotors,
blue, AC/DC input 230 V AC, EuroPlug
Expand
View Pricing
Z742581
Mini-Centrifuge with 2 rotors,
red, AC/DC input 230 V AC, UK plug
Expand
View Pricing
Z742584
Mini-Centrifuge with 2 rotors,
blue, AC/DC input 230 V AC, UK plug
Expand
View Pricing
Z742580
Mini-Centrifuge with 2 rotors,
red, AC/DC input 230 V AC, EuroPlug
Expand
View Pricing
Z742579
Mini-Centrifuge with 2 rotors,
red, AC/DC input 115 V AC, US 3-pin plug
Expand
View Pricing