Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microflex
®
Dura Flock
®
nitrile gloves
Products
Product Category
gloves (5)
Manufacturer
Length (in)
Color
Package Size
Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z675822
Microflex
®
Dura Flock
®
nitrile gloves,
size M
Expand
View Pricing
Z675830
Microflex
®
Dura Flock
®
nitrile gloves,
size L
Expand
View Pricing
Z675849
Microflex
®
Dura Flock
®
nitrile gloves,
size XL
Expand
View Pricing
Z675814
Microflex
®
Dura Flock
®
nitrile gloves,
size S
Expand
View Pricing
Z675857
Microflex
®
Dura Flock
®
nitrile gloves,
size XXL
Expand
View Pricing