
Methyl p-toluate

Synonym(s):
(Methyl 4-methylbenzoate), Methyl p-toluate
Linear Formula:
CH3C6H4CO2CH3
CAS Number:
Molecular Weight:
150.17
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing