Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium stearate
Synonym(s):
Stearic acid magnesium salt
Linear Formula:
[CH
3
(CH
2
)
16
CO
2
]
2
Mg
CAS Number:
557-04-0
Molecular Weight:
591.24
EC Number:
209-150-3
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Application
Markush Class
Markush Group
Special Grade
Puriss (1)
Technical (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
415057
Magnesium stearate,
technical grade
Expand
View Pricing
26454
Magnesium stearate,
puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Expand
View Pricing