Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium hydroxide
Linear Formula:
Mg(OH)
2
CAS Number:
1309-42-8
Molecular Weight:
58.32
EC Number:
215-170-3
Products
Product Category
salts (2)
inorganic bases (1)
Brand
Sigma-Aldrich (3)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
310093
Magnesium hydroxide,
reagent grade, 95%
Expand
View Pricing
63081
Magnesium hydroxide,
BioUltra, ≥99.0% (KT)
Expand
View Pricing
1374260
Magnesium hydroxide,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
19-0280
Magnesium hydroxide,
SAJ first grade, ≥95.0%
Expand
View Pricing