Skip to Content
Merck
L-Lysine monohydrochloride SAJ special grade, ≥99.0%

L-Lysine monohydrochloride

L-Lysine monohydrochloride SAJ special grade, ≥99.0%
Synonym(s):
(S)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride
Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
Reference Material Type
Special Grade
Technique