Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium sulfate monohydrate
Linear Formula:
Li
2
SO
4
· H
2
O
CAS Number:
10102-25-7
Molecular Weight:
127.96
EC Number:
233-820-4
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (3)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
398152
Lithium sulfate monohydrate,
ACS reagent, ≥99.0% dry basis
Expand
View Pricing
62612
Lithium sulfate monohydrate,
BioUltra, ≥99.0% (T)
Expand
View Pricing
62609
Lithium sulfate monohydrate,
puriss. p.a., ACS reagent, ≥99.0% (dried basis, T)
Expand
View Pricing