Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium sulfate
Linear Formula:
Li
2
SO
4
CAS Number:
10377-48-7
Molecular Weight:
109.94
EC Number:
233-820-4
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (4)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Anhydrous (1)
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
920339
Lithium sulfate,
anhydrous, 99.5% trace metals basis
Expand
View Pricing
L6375
Lithium sulfate,
≥98.5% (titration)
Expand
View Pricing
203653
Lithium sulfate,
≥99.99% trace metals basis
Expand
View Pricing
62613
Lithium sulfate,
purum p.a., ≥98.0% (T)
Expand
View Pricing