Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lipoxidase from
Glycine max
(soybean)
Synonym(s):
Linoleate:oxygen oxidoreductase, Lipoxygenase
CAS Number:
9029-60-1
EC Number:
232-853-1
Products
Product Category
enzymes (1)
peptides (1)
Brand
Sigma-Aldrich (2)
Physical Form
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
L7395
Lipoxidase from
Glycine max
(soybean),
Type I-B, lyophilized powder, ≥50,000 units/mg solid
Expand
View Pricing
L6632
Lipoxidase from
Glycine max
(soybean),
Type V, ammonium sulfate suspension, 500,000-1,000,000 units/mg protein
Expand
View Pricing