
L-(+)-Lactic acid solution

Synonym(s):
(S)-(+)-Lactic acid, (S)-2-Hydroxypropionic acid, Sarcolactic acid
Linear Formula:
CH3CH(OH)CO2H
CAS Number:
Molecular Weight:
90.08
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Optical Isomer
Special Grade
Showing 1-4 of 4