Skip to Content
Merck
Lauroyl coenzyme A lithium salt ≥90% (HPLC)

Lauroyl coenzyme A lithium salt

Lauroyl coenzyme A lithium salt ≥90% (HPLC)
Synonym(s):
C12-CoA, Dodecanoyl-Coenzyme A lithium salt
Linear Formula:
C33H58N7O17P3S
CAS Number:
Molecular Weight:
955.77

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing