Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lactoferrin from human milk
CAS Number:
936541-36-5
Products
Product Category
proteins (4)
media supplements (1)
Brand
Sigma-Aldrich (5)
Biological Source
Color
Gene Alias
Physical Form
Product Line
Purity
Tissue
Chemical Composition
Quality Segment
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
L3770
Lactoferrin from human milk,
Iron saturated, ≥85% (SDS-PAGE)
Expand
View Pricing
L0520
Lactoferrin from human milk,
≥85% (SDS-PAGE), lyophilized powder
Expand
View Pricing
L4894
Lactoferrin from human milk,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
61327
Lactoferrin from human milk,
iron-saturated, ≥85% (GE)
Expand
View Pricing
61326
Lactoferrin from human milk,
~80% (GE)
Expand
View Pricing